siRNA Details
| Gene Symbol: CSAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129826.2 | NP_001123298.1 | 2 | 820 | ||
| NM_001129828.2 | NP_001123300.1 | 2 | 769 | |||
| NR_130638.1 | 5 | 762 | ||||
| GenBank mRNA | BC007228.1 | - | 5 | 762 | ||
| Gene Symbol: CSAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724857.3 | XP_006724920.1 | 2 | 946 | ||
| XM_017030023.1 | XP_016885512.1 | 2 | 895 | |||

CSAG3
Genomic Map