siRNA Details
| Gene Symbol: PAGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207339.3 | NP_997222.1 | 3 | 243 | ||
| XM_017029353.1 | XP_016884842.1 | 3 | 272 | |||
| GenBank mRNA | BC054022.1 | - | 3 | 237 | 6603202 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAGE2| Gene Symbol: PAGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207339.3 | NP_997222.1 | 3 | 243 | ||
| XM_017029353.1 | XP_016884842.1 | 3 | 272 | |||
| GenBank mRNA | BC054022.1 | - | 3 | 237 | 6603202 | |