siRNA Details
| Gene Symbol: GRINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000837.1 | NP_000828.1 | 7 | 1651 | ||
| NM_001009184.1 | NP_001009184.1 | 7 | 1539 | |||
| GenBank mRNA | AB097026.1 | - | 7 | 1463 | ||
| AK127640.1 | - | 7 | 1687 | |||
| AL157442.1 | - | 1 | 382 | |||
| BC041788.1 | - | 7 | 1497 | 4738372 | ||
| BC065834.1 | - | 6 | 1405 | |||
| BC084553.1 | - | 6 | 1388 | 6206624 | ||

GRINA
Genomic Map