siRNA Details
| Gene Symbol: GRINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000837.1 | NP_000828.1 | 3 | 673 | ||
| NM_001009184.1 | NP_001009184.1 | 3 | 561 | |||
| GenBank mRNA | AB097026.1 | - | 3 | 485 | ||
| AK127640.1 | - | 3 | 709 | |||
| AK294127.1 | - | 4 | 418 | |||
| BC041788.1 | - | 3 | 519 | 4738372 | ||
| BC065834.1 | - | 2 | 427 | |||
| BC084553.1 | - | 2 | 410 | 6206624 | ||
| BM762447.1 | - | 3 | 591 | |||

GRINA
Genomic Map