siRNA Details
| Gene Symbol: DEFB108B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002035.1 | NP_001002035.1 | 1 | 13 | ||
| GenBank mRNA | AF529416.1 | - | 1 | 13 | ||
| Gene Symbol: LOC100133128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008898.1 | XP_016864387.1 | 1 | 189 | ||

DEFB108B
Genomic Map