siRNA Details
| Gene Symbol: DEFB108B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002035.1 | NP_001002035.1 | 2 | 119 | ||
| GenBank mRNA | AF529416.1 | - | 2 | 119 | ||
| AY122468.1 | - | 1 | 56 | |||
| BX095678.1 | - | 3 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEFB108B| Gene Symbol: DEFB108B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002035.1 | NP_001002035.1 | 2 | 119 | ||
| GenBank mRNA | AF529416.1 | - | 2 | 119 | ||
| AY122468.1 | - | 1 | 56 | |||
| BX095678.1 | - | 3 | 356 | |||