siRNA Details
| Gene Symbol: SERTAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019605.3 | NP_062551.1 | 3 | 423 | ||
| XM_005273193.4 | XP_005273250.1 | 3 | 623 | |||
| GenBank mRNA | AK096381.1 | - | 3 | 389 | ||
| AK315386.1 | - | 2 | 422 | |||
| BC012083.1 | - | 3 | 423 | 4564813 | ||

SERTAD4
Genomic Map