siRNA Details
| Gene Symbol: OR5L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004738.1 | NP_001004738.1 | 1 | 859 | ||
| GenBank mRNA | BC109249.1 | - | 1 | 859 | 40022949 | |
| BC109250.1 | - | 1 | 859 | 40022959 | ||
| BC136939.1 | - | 1 | 959 | 9020931 | ||

OR5L1
Genomic Map