siRNA Details
| Gene Symbol: OR9G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005213.1 | NP_001005213.1 | 1 | 177 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR9G1| Gene Symbol: OR9G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005213.1 | NP_001005213.1 | 1 | 177 | ||
Set Membership: |
|