siRNA Details
| Gene Symbol: PAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017931.2 | NP_001017931.2 | 4 | 525 | ||
| NM_001171252.1 | NP_001164723.1 | 4 | 539 | |||
| NM_001303613.1 | NP_001290542.1 | 4 | 327 | |||
| XM_017029282.1 | XP_016884771.1 | 4 | 328 | |||
| GenBank mRNA | BC140001.1 | - | 4 | 398 | 8860308 | |
| BC140741.1 | - | 4 | 411 | 9021609 | ||
| BC144449.1 | - | 4 | 424 | 9052975 | ||
| HY049011.1 | - | 4 | 327 | |||

PAGE3
Genomic Map