siRNA Details
| Gene Symbol: FAM78B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017961.4 | NP_001017961.1 | 2 | 913 | ||
| NR_135199.1 | 2 | 913 | ||||
| XM_017000417.1 | XP_016855906.1 | 2 | 913 | |||
| XM_017000418.1 | XP_016855907.1 | 2 | 464 | |||
| GenBank mRNA | AK299946.1 | - | 2 | 443 | ||
| BC114214.1 | - | 3 | 996 | 5588830 | ||

FAM78B
Genomic Map