siRNA Details
| Gene Symbol: ROPN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012337.2 | NP_001012337.1 | 6 | 900 | ||
| NM_001308313.1 | NP_001295242.1 | 7 | 946 | |||
| XM_005247138.3 | XP_005247195.1 | 5 | 755 | |||
| XM_006713513.3 | XP_006713576.1 | 8 | 1042 | |||
| GenBank mRNA | AF231410.1 | - | 6 | 859 | ||
| BC015413.1 | - | 4 | 640 | 4394326 | ||
| BC141849.1 | - | 7 | 893 | 8860347 | ||

ROPN1B
Genomic Map