siRNA Details
| Gene Symbol: METTL21C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010977.2 | NP_001010977.1 | 4 | 515 | ||
| XM_017020407.1 | XP_016875896.1 | 5 | 1409 | |||
| XM_017020408.1 | XP_016875897.1 | 5 | 1962 | |||
| GenBank mRNA | BC132748.1 | - | 4 | 515 | 40146770 | |
| BC132750.1 | - | 4 | 515 | 40146772 | ||
| BX089441.1 | - | 1 | 83 | |||

METTL21C
Genomic Map