siRNA Details
| Gene Symbol: NUDT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012758.2 | NP_001012776.1 | 7 | 861 | ||
| XM_011509260.2 | XP_011507562.1 | 5 | 1299 | |||
| GenBank mRNA | AA287243.1 | - | 2 | 193 | ||
| AK091752.1 | - | 3 | 2120 | |||
| AK298083.1 | - | 5 | 1393 | |||
| BC046352.1 | - | 5 | 504 | |||

NUDT17
Genomic Map