siRNA Details
| Gene Symbol: ALG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015050.2 | NP_001015050.2 | 6 | 647 | ||
| NM_001195223.1 | NP_001182152.1 | 7 | 758 | |||
| XM_017005844.1 | XP_016861333.1 | 7 | 4025 | |||
| XM_017005845.1 | XP_016861334.1 | 7 | 651 | |||
| XM_017005846.1 | XP_016861335.1 | 6 | 3914 | |||
| XM_017005847.1 | XP_016861336.1 | 6 | 540 | |||
| GenBank mRNA | BC073816.1 | - | 6 | 642 | 5419254 | |

ALG1L
Genomic Map