siRNA Details
| Gene Symbol: PLAC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012973.1 | NP_001012991.1 | 2 | 408 | ||
| GenBank mRNA | BC066348.1 | - | 2 | 408 | 30339526 | |
| BC090922.1 | - | 2 | 408 | 30331098 | ||
| BC117332.1 | - | 2 | 236 | 40125883 | ||
| BC117334.1 | - | 2 | 236 | 40125885 | ||

PLAC9
Genomic Map