siRNA Details
| Gene Symbol: LRRC73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012974.2 | NP_001012992.1 | 6 | 1453 | ||
| NM_001271882.1 | NP_001258811.1 | 6 | 761 | |||
| GenBank mRNA | AK126786.1 | - | 5 | 673 | ||
| BC107130.2 | - | 6 | 661 | |||
| BC108661.1 | - | 6 | 1453 | 5240367 | ||

LRRC73
Genomic Map