siRNA Details
| Gene Symbol: TSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010873.2 | NP_001010873.1 | 3 | 584 | ||
| NM_001159726.1 | NP_001153198.1 | 3 | 394 | |||
| XM_011514396.2 | XP_011512698.1 | 3 | 387 | |||
| XM_011514397.2 | XP_011512699.1 | 3 | 916 | |||
| GenBank mRNA | BC137560.1 | - | 3 | 364 | 9021560 | |
| BC137565.1 | - | 3 | 364 | 9021565 | ||
| BC144710.1 | - | 3 | 394 | 9053248 | ||
| BU655765.1 | - | 3 | 594 | |||
| BU657409.1 | - | 3 | 569 | |||

TSPO2
Genomic Map