siRNA Details
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 24 | 3653 | ||
NM_001162501.1 | NP_001155973.1 | 23 | 5974 | |||
NM_015088.2 | NP_055903.2 | 21 | 5644 | |||
GenBank mRNA | AK294519.1 | - | 23 | 5965 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144812, NM_177124, AK052247, AK122440, AK162788, and others.
Panther Classification:
Gene Ontology Categories:
