siRNA Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303450.1 | NP_001290379.1 | 7 | 1965 | ||
| NM_021646.3 | NP_067678.1 | 6 | 2828 | |||
| XM_005255243.3 | XP_005255300.1 | 5 | 2794 | |||
| XM_011522454.2 | XP_011520756.1 | 5 | 2583 | |||
| XM_017023121.1 | XP_016878610.1 | 5 | 2321 | |||
| GenBank mRNA | AB011129.1 | - | 5 | 2568 | ||
| AF007129.1 | - | 1 | 1083 | |||
| AK297993.1 | - | 7 | 1964 | |||
| BC004125.2 | - | 1 | 1769 | |||
| BC033151.1 | - | 1 | 1898 | |||

ZNF500
Genomic Map