siRNA Details
| Gene Symbol: CEP170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112726.2 | NP_001106197.1 | 17, 18 | 4601 | ||
| NM_015005.2 | NP_055820.2 | 16, 17 | 4574 | |||
| XM_005267550.3 | XP_005267607.1 | 17, 18 | 4688 | |||
| XM_011536665.2 | XP_011534967.1 | 18, 19 | 4841 | |||
| XM_011536666.2 | XP_011534968.1 | 17, 18 | 5204 | |||
| XM_011536667.2 | XP_011534969.1 | 10, 11 | 2880 | |||
| XM_017021227.1 | XP_016876716.1 | 16, 17 | 5453 | |||
| XM_017021228.1 | XP_016876717.1 | 16, 17 | 5348 | |||
| GenBank mRNA | AB006622.3 | - | 17, 18 | 4601 | ||
| AK025023.1 | - | 2, 3 | 176 | |||
| AK091980.1 | - | 5, 6 | 2210 | |||
| AK300834.1 | - | 3, 4 | 362 | |||
| BC047913.1 | - | 3, 4 | 269 | |||
| BC112928.1 | - | 16, 17 | 4605 | 6141521 | ||

CEP170B
Genomic Map