siRNA Details
| Gene Symbol: C16orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272051.1 | NP_001258980.1 | 2 | 698 | ||
| GenBank mRNA | AB101207.1 | - | 2 | 642 | ||
| BC092497.1 | - | 2 | 642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf91| Gene Symbol: C16orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272051.1 | NP_001258980.1 | 2 | 698 | ||
| GenBank mRNA | AB101207.1 | - | 2 | 642 | ||
| BC092497.1 | - | 2 | 642 | |||