siRNA Details
| Gene Symbol: ZNF763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012753.1 | NP_001012771.1 | 4 | 1130 | ||
| GenBank mRNA | AK092240.1 | - | 4 | 1130 | ||
| AK299182.1 | - | 4 | 1162 | |||
| AK299224.1 | - | 2 | 933 | |||
| BC105711.1 | - | 4 | 1093 | 40024621 | ||
| BC105712.1 | - | 4 | 1093 | 40024618 | ||
| BC105713.1 | - | 4 | 1093 | 40024617 | ||

ZNF763
Genomic Map