siRNA Details
| Gene Symbol: CFAP157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518559.2 | XP_011516861.2 | 8 | 2802 | ||
| GenBank mRNA | AK126437.1 | - | 4 | 2106 | ||
| AL833241.1 | - | 2 | 2686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP157| Gene Symbol: CFAP157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518559.2 | XP_011516861.2 | 8 | 2802 | ||
| GenBank mRNA | AK126437.1 | - | 4 | 2106 | ||
| AL833241.1 | - | 2 | 2686 | |||