siRNA Details
| Gene Symbol: ERO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019891.3 | NP_063944.3 | 1, 2 | 366 | ||
| XM_017001834.1 | XP_016857323.1 | 1, 2 | 285 | |||
| GenBank mRNA | AF252538.1 | - | 1, 2 | 283 | ||
| AK293941.1 | - | 1, 2 | 347 | |||
| AK315351.1 | - | 1, 2 | 285 | |||
| BC032823.2 | - | 1, 2 | 217 | 4829502 | ||
| BC044573.1 | - | 1, 2 | 312 | 5270105 | ||
| BG723126.1 | - | 1, 2 | 369 | |||

ERO1B
Genomic Map