siRNA Details
| Gene Symbol: LINC00544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033889.1 | 5 | 594 | |||
| GenBank mRNA | BC132810.1 | - | 3 | 325 | ||
| BC132836.1 | - | 3 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00544| Gene Symbol: LINC00544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033889.1 | 5 | 594 | |||
| GenBank mRNA | BC132810.1 | - | 3 | 325 | ||
| BC132836.1 | - | 3 | 325 | |||