siRNA Details
| Gene Symbol: LOC440570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135765.1 | 1 | 1023 | |||
| GenBank mRNA | AK125737.1 | - | 1 | 1024 | ||
| BC126340.1 | - | 1 | 208 | 8992056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440570| Gene Symbol: LOC440570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135765.1 | 1 | 1023 | |||
| GenBank mRNA | AK125737.1 | - | 1 | 1024 | ||
| BC126340.1 | - | 1 | 208 | 8992056 | ||