siRNA Details
| Gene Symbol: LINC00969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128346.1 | - | 2 | 2303 | ||
| BC144457.1 | - | 2 | 1834 | |||
| KJ535002.1 | - | 2 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00969| Gene Symbol: LINC00969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128346.1 | - | 2 | 2303 | ||
| BC144457.1 | - | 2 | 1834 | |||
| KJ535002.1 | - | 2 | 424 | |||