siRNA Details
| Gene Symbol: ANKRD20A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012419.2 | NP_001012419.1 | 15 | 3748 | ||
| Gene Symbol: ANKRD20A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR936649.1 | - | 2 | 2802 | ||

ANKRD20A3
Genomic Map