siRNA Details
| Gene Symbol: ANKRD18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014576.1 | XP_016870065.1 | 7, 8 | 1156 | ||
| XM_017014577.1 | XP_016870066.1 | 7, 8 | 1149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD18A| Gene Symbol: ANKRD18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014576.1 | XP_016870065.1 | 7, 8 | 1156 | ||
| XM_017014577.1 | XP_016870066.1 | 7, 8 | 1149 | |||