siRNA Details
| Gene Symbol: SPANXN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009616.3 | NP_001009616.1 | 2 | 530 | ||
| GenBank mRNA | AL039107.1 | - | 1 | 126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPANXN5| Gene Symbol: SPANXN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009616.3 | NP_001009616.1 | 2 | 530 | ||
| GenBank mRNA | AL039107.1 | - | 1 | 126 | ||