siRNA Details
| Gene Symbol: SPRR2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006945.4 | NP_008876.3 | 2 | 374 | ||
| XM_017002175.1 | XP_016857664.1 | 2 | 406 | |||
| GenBank mRNA | AK291317.1 | - | 2 | 376 | ||
| BG183546.1 | - | 3 | 603 | |||
| M21302.1 | - | 2 | 374 | |||

SPRR2D
Genomic Map