siRNA Details
| Gene Symbol: SPRR2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 370 | ||
| XM_017002177.1 | XP_016857666.1 | 4 | 747 | |||
| XM_017002178.1 | XP_016857667.1 | 2 | 293 | |||
| GenBank mRNA | AA490630.1 | - | 1 | 280 | ||
| BC130455.1 | - | 2 | 370 | |||
| BC133041.1 | - | 2 | 370 | |||

SPRR2G
Genomic Map