siRNA Details
| Gene Symbol: CIB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029881.2 | NP_001025052.1 | 4 | 294 | ||
| XM_011532514.2 | XP_011530816.1 | 4 | 377 | |||
| XM_011532517.2 | XP_011530819.1 | 2 | 664 | |||
| XM_017003329.1 | XP_016858818.1 | 3 | 1148 | |||
| XM_017003330.1 | XP_016858819.1 | 2 | 1058 | |||
| GenBank mRNA | BC127684.1 | - | 4 | 294 | 40127392 | |
| BC127685.1 | - | 4 | 292 | 40127393 | ||
| BC140923.1 | - | 4 | 268 | 9056735 | ||

CIB4
Genomic Map