siRNA Details
| Gene Symbol: SLC35F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029858.3 | NP_001025029.2 | 3 | 626 | ||
| XM_005266865.4 | XP_005266922.1 | 3 | 926 | |||
| GenBank mRNA | AK295860.1 | - | 3 | 323 | ||
| BC028615.1 | - | 3 | 629 | 4817165 | ||
| BC036493.1 | - | 3 | 395 | |||
| BC114925.1 | - | 4 | 500 | 40025808 | ||

SLC35F1
Genomic Map