siRNA Details
| Gene Symbol: SIPA1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015073.2 | NP_055888.1 | 3 | 877 | ||
| XM_005258671.4 | XP_005258728.1 | 3 | 1318 | |||
| XM_011526657.2 | XP_011524959.1 | 3 | 1672 | |||
| XM_017026517.1 | XP_016882006.1 | 4 | 1772 | |||
| XM_017026518.1 | XP_016882007.1 | 3 | 1274 | |||
| GenBank mRNA | AY168880.1 | - | 1 | 623 | ||
| BC036863.1 | - | 3 | 849 | |||
| BC150620.1 | - | 3 | 856 | 9056990 | ||

SIPA1L3
Genomic Map