siRNA Details
| Gene Symbol: LINC01366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026945.2 | 3 | 696 | |||
| NR_108024.1 | 3 | 677 | ||||
| NR_108025.1 | 2 | 606 | ||||
| GenBank mRNA | AK093725.1 | - | 3 | 1392 | ||
| BC093717.1 | - | 1 | 169 | |||
| BC111994.1 | - | 1 | 169 | |||
| BQ053612.1 | - | 3 | 658 | |||
| CD518314.1 | - | 3 | 641 | |||

LINC01366
Genomic Map