siRNA Details
| Gene Symbol: LINC01366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026945.2 | 3 | 843 | |||
| NR_108024.1 | 3 | 824 | ||||
| NR_108025.1 | 2 | 753 | ||||
| GenBank mRNA | AK093725.1 | - | 3 | 1539 | ||
| BC093717.1 | - | 1 | 316 | |||
| BC111994.1 | - | 1 | 316 | |||

LINC01366
Genomic Map