siRNA Details
| Gene Symbol: TM9SF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020123.3 | NP_064508.3 | 2 | 342 | ||
| XM_011539976.2 | XP_011538278.1 | 2 | 422 | |||
| XM_011539977.1 | XP_011538279.1 | 2 | 422 | |||
| GenBank mRNA | AF116347.1 | - | 2 | 92 | ||
| AF160213.1 | - | 2 | 141 | |||
| AF269150.1 | - | 2 | 342 | |||
| AK314348.1 | - | 2 | 308 | |||
| AY358819.1 | - | 11 | ||||
| BC136788.1 | - | 2 | 342 | 9020778 | ||
| BC136789.1 | - | 2 | 342 | 9020779 | ||

TM9SF3
Genomic Map