siRNA Details
| Gene Symbol: KCTD21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029859.2 | NP_001025030.1 | 2 | 1717 | ||
| XM_005273925.3 | XP_005273982.1 | 2 | 1942 | |||
| XM_005273928.1 | XP_005273985.1 | 3 | 1842 | |||
| XM_006718516.3 | XP_006718579.1 | 2 | 4887 | |||
| XM_006718517.2 | XP_006718580.1 | 3 | 1954 | |||
| XM_006718518.3 | XP_006718581.1 | 2 | 1702 | |||
| XM_011544955.2 | XP_011543257.1 | 2 | 3182 | |||
| GenBank mRNA | AK095233.1 | - | 3 | 1466 | ||
| BC036058.1 | - | 2 | 1706 | 5277643 | ||

KCTD21
Genomic Map