siRNA Details
| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026836.1 | 2 | 844 | |||
| NR_026837.1 | 1 | 1446 | ||||
| GenBank mRNA | AK095156.1 | - | 1 | 463 | ||
| BC040230.1 | - | 2 | 819 | |||
| BC093901.1 | - | 1 | 372 | |||
| BC093903.1 | - | 1 | 372 | |||
| DA139014.1 | - | 1 | 502 | |||

TRHDE-AS1
Genomic Map