siRNA Details
| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026837.1 | 1 | 1308 | |||
| GenBank mRNA | AK095156.1 | - | 1 | 325 | ||
| BC093901.1 | - | 1 | 234 | |||
| BC093903.1 | - | 1 | 234 | |||
| DA139014.1 | - | 1 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRHDE-AS1| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026837.1 | 1 | 1308 | |||
| GenBank mRNA | AK095156.1 | - | 1 | 325 | ||
| BC093901.1 | - | 1 | 234 | |||
| BC093903.1 | - | 1 | 234 | |||
| DA139014.1 | - | 1 | 364 | |||