siRNA Details
| Gene Symbol: LOC338797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103736.1 | 3 | 728 | |||
| GenBank mRNA | AK098523.1 | - | 3 | 728 | ||
| BC093897.1 | - | 1 | 256 | 7939742 | ||
| BC093899.1 | - | 1 | 256 | 7939744 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC338797| Gene Symbol: LOC338797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103736.1 | 3 | 728 | |||
| GenBank mRNA | AK098523.1 | - | 3 | 728 | ||
| BC093897.1 | - | 1 | 256 | 7939742 | ||
| BC093899.1 | - | 1 | 256 | 7939744 | ||