siRNA Details
| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029871.3 | NP_001025042.2 | 5 | 2419 | ||
| NM_001040007.2 | NP_001035096.1 | 4 | 2233 | |||
| GenBank mRNA | AK122609.1 | - | 5 | 2415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPO4| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029871.3 | NP_001025042.2 | 5 | 2419 | ||
| NM_001040007.2 | NP_001035096.1 | 4 | 2233 | |||
| GenBank mRNA | AK122609.1 | - | 5 | 2415 | ||