siRNA Details
| Gene Symbol: SOWAHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029870.1 | NP_001025041.1 | 1 | 232 | ||
| GenBank mRNA | BC137241.1 | - | 1 | 309 | 9021238 | |
| BC137255.1 | - | 1 | 309 | 9021252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOWAHB| Gene Symbol: SOWAHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029870.1 | NP_001025041.1 | 1 | 232 | ||
| GenBank mRNA | BC137241.1 | - | 1 | 309 | 9021238 | |
| BC137255.1 | - | 1 | 309 | 9021252 | ||