siRNA Details
| Gene Symbol: C6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029863.2 | NP_001025034.1 | 1 | 1981 | ||
| NM_001317342.1 | NP_001304271.1 | 2 | 1741 | |||
| GenBank mRNA | AF055030.1 | - | 1 | 1004 | ||
| AF451991.1 | - | 1 | 1926 | |||
| BC051700.1 | - | 1 | 1932 | 4792634 | ||

C6orf120
Genomic Map