siRNA Details
| Gene Symbol: C6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029863.2 | NP_001025034.1 | 1 | 3830 | ||
| NM_001317342.1 | NP_001304271.1 | 2 | 3590 | |||
| GenBank mRNA | BC051700.1 | - | 1 | 3781 | 4792634 | |
| CR735892.1 | - | 1 | 455 | |||

C6orf120
Genomic Map