siRNA Details
| Gene Symbol: REP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029874.1 | NP_001025045.1 | 1 | 484 | ||
| GenBank mRNA | AY662682.1 | - | 1 | 419 | ||
| BC140921.1 | - | 1 | 460 | 9056733 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

REP15| Gene Symbol: REP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029874.1 | NP_001025045.1 | 1 | 484 | ||
| GenBank mRNA | AY662682.1 | - | 1 | 419 | ||
| BC140921.1 | - | 1 | 460 | 9056733 | ||