siRNA Details
| Gene Symbol: RGS7BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029875.2 | NP_001025046.1 | 6 | 1375 | ||
| NM_001271890.1 | NP_001258819.1 | 4 | 489 | |||
| NM_001271891.1 | NP_001258820.1 | 4 | 487 | |||
| NR_073511.1 | 5 | 574 | ||||
| GenBank mRNA | AK294808.1 | - | 5 | 480 | ||
| AK296443.1 | - | 6 | 1023 | |||
| AK296494.1 | - | 4 | 457 | |||
| BC140707.1 | - | 6 | 721 | 9021575 | ||
| BX640900.1 | - | 6 | 613 | |||
| DA417118.1 | - | 5 | 541 | |||
| DC342795.1 | - | 4 | 440 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_029879, BC118957, AK028323, AK043529, AK137417, and others.

RGS7BP
Genomic Map