siRNA Details
Gene Symbol: LOC441178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117092.1 | 1 | 35 | |||
GenBank mRNA | AK056600.1 | - | 1 | 35 | ||
AL832737.1 | - | 4 | 797 | |||
BC093745.1 | - | 1 | 304 | 7939590 | ||
BC111990.1 | - | 1 | 305 | 8327458 |
